Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KDM1B, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89113558 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-558 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-558 Supplier Abnova Corporation Supplier No. MAB8742
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant KDM1B.

Specifications

Antigen KDM1B
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant KDM1B.
Dilution Western Blot (1:100-1:2000) ELISA (1:5000-1:20000) The optimal working dilution should be determined by the end user.
Formulation In 100mM Tris-Glycine, pH 7.4, 150mM NaCl (50% glycerol, 1% BSA, 0.02% sodium azide)
Gene AOF1
Gene Accession No. NM_153042
Gene Alias C6orf193/DKFZp686I0412/FLJ33898/FLJ34109/FLJ43328/bA204B7.3/dJ298J15.2
Gene Symbols AOF1
Host Species Mouse
Immunogen Recombinant protein corresponding to C-terminus residues of human KDM1B.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221656
Test Specificity This antibody is specific to KDM1B.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.