Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KEL, Mouse, Clone: BRIC203, Abnova™

Catalog No. 89112189 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-189 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-189 Supplier Abnova Corporation Supplier No. MAB5250
Add to Cart
Add to Cart
Request a Quote

Mouse monoclonal antibody raised against native KEL.

This gene encodes a type II transmembrane glycoprotein that is the highly polymorphic Kell blood group antigen. The Kell glycoprotein links via a single disulfide bond to the XK membrane protein that carries the Kx antigen. The encoded protein contains sequence and structural similarity to members of the neprilysin (M13) family of zinc endopeptidases. [provided by RefSeq

Specifications

Antigen KEL
Applications Agglutination, Immunoprecipitation
Classification Monoclonal
Clone BRIC203
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native KEL.
Dilution The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, pH 7.4 (0.09% sodium azide)
Gene KEL
Gene Alias CD238/ECE3
Gene Symbols KEL
Host Species Mouse
Immunogen Native purified KEL from human erythrocytes.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3792
Test Specificity This antibody recognizes human CD238, also known as the kell blood group antigen Kpbc. The kell blood group antigens are carried on a 93kD erythrocyte membrane glycoprotein. In addition clone BRIC203 will agglutinate Kp (a+b-c+) and Kp (a-b-c+) erythrocytes but does not react with Ko erythrocytes.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.