Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KHK (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89104807 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-807 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-807 Supplier Abnova Corporation Supplier No. H00003795A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant KHK.

This gene encodes ketohexokinase that catalyzes conversion of fructose to fructose-1-phosphate. The product of this gene is the first enzyme with a specialized pathway that catabolizes dietary fructose. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Sequence: MEEKQILCVGLVVLDVISLVDKYPKEDSEIRCLSQRWQRGGNASNSCTILSLLGAPCAFMGSMAPGHVADFVLDDLRRYSVDLRYTVFQTTGSVPIATVIINEASGSRTILYYDRSLPDVSATDFEKVDLTQFKWIHIEGRNASEQVKMLQRIDAHNTRQPPEQKIRVSVEVEKPREELFQLFGYGDVVFVSKDVAKHLGFQSAEEALRGLYGRVRKGAVLVCAWAEEGADALGPDGKLLHSDAFPPPRVVDTLGAGDTFNASVIFSLSQGRSVQEALRFGCQVAGKKCGLQGFDGIV

Specifications

Antigen KHK
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant KHK.
Formulation 50% glycerol
Gene KHK
Gene Accession No. BC006233
Gene Symbols KHK
Host Species Mouse
Immunogen KHK (AAH06233, 1 a.a. ∽ 298 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3795
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.