Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIF11, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89113536 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-536 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-536 Supplier Abnova Corporation Supplier No. MAB8717
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant KIF11.

This gene encodes a motor protein that belongs to the kinesin-like protein family. Members of this protein family are known to be involved in various kinds of spindle dynamics. The function of this gene product includes chromosome positioning, centrosome separation and establishing a bipolar spindle during cell mitosis. [provided by RefSeq

Specifications

Antigen KIF11
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant KIF11.
Dilution Western Blot (1:100-1:2000) ELISA (1:5000-1:20000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide, 1% BSA, 50% glycerol)
Gene KIF11
Gene Accession No. NM_004523
Gene Alias EG5/HKSP/KNSL1/TRIP5
Gene Symbols KIF11
Host Species Mouse
Immunogen Recombinant protein corresponding to N-terminus residues of human KIF11.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3832
Test Specificity This antibody is specific to KIF11.
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.