Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-KIF3A, Clone: 4F9.1,

Catalog No. MABC155MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC155MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC155MI Supplier MilliporeSigma™ Supplier No. MABC155
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects KIF3A clone: 4F9.1 in Human, Mouse, Rat samples, and it is validated for Immunohistochemistry, Western Blotting

Kinesin-like protein KIF3A (UniProt Q9Y496; also known as Kinesin family protein 3A, Microtubule plus end-directed kinesin motor 3A) is encoded by the KIF3A (also known as FLA10, KIF3, KLP-20) gene (Gene ID 11127) in human. Kinesin-2 is a heterotrimeric motor protein composed of two kinesin-related subunits, KIF3A and KIF3B, and an associated protein KAP3. The kinesin-2 motor KIF3A/B drives intraflagellar transport (IFT) of tubulins and other axonemal building blocks in cilia and flagella, pigment granules in melanosomes, and vesicles in neurons. Defects in these transport processes are linked to polycystic kidney disease and neurodegenerative diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIF3A
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4F9.1
Formulation Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. Q9Y496
Gene Symbols KIF3; KIF3A
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to the C-terminal of human KIF3A.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_001287720
Target Species Human, Mouse, Rat
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.