Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kinesin (heavy chain), Mouse, Clone: KN-02, Abnova™

Catalog No. 89135480 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-480 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-480 Supplier Abnova Corporation Supplier No. MAB3621
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Kinesin (heavy chain).

Specifications

Antigen Kinesin (heavy chain)
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone KN-02
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Kinesin (heavy chain).
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified porcine Kinesin (heavy chain).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody recognizes heavy chain of conventional kinesin associated withvesicles and with lower affinity with denaturated molecule. Epitope is located in coiled-coilstalk domain. It stains Western blots of kinesin-enriched preparations. Epitope mapping (by limited proteolysis of partially purified porcine kinesin) followed by immunoblotting has revealed that antibodies MAB3620, MAB3621 and MAB3622 react with differentsets of fragments. This antibody does not react with kinesin bound to taxol-stabilized microtubules.
Target Species Human, Mouse, Pig, Rat
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.