Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kinesin (heavy chain), Mouse, Clone: KN-03, Abnova™
Mouse monoclonal antibody raised against native Kinesin (heavy chain).
Supplier: Abnova Corporation MAB3622
Specifications
Kinesin (heavy chain) | |
Monoclonal | |
1 mg/mL | |
Mouse monoclonal antibody raised against native Kinesin (heavy chain). | |
In PBS, pH 7.4 (0.09% sodium azide) | |
Native purified porcine Kinesin (heavy chain). | |
RUO | |
This antibody recognizes heavy chain of conventional kinesin associated with vesicles and with lower affinity with denaturated molecule. It stains Western blots ofkinesin-enriched preparations. Epitope mapping (by limited proteolysis of partially purified porcine kinesin) followed by immunoblotting has revealed that antibodies MAB3620, MAB3621 and MAB3622 react with differentsets of fragments. This antibody well recognizes kinesin bound to taxol-stabilized microtubules. | |
Store at 4°C. Do not freeze. | |
IgM |
Immunocytochemistry, Immunofluorescence | |
KN-03 | |
Unconjugated | |
The optimal working dilution should be determined by the end user. | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse, Pig, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction