Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KITLG, Mouse, Clone: four helix cytokine core, Abnova™

Catalog No. 89113178 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-178 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-178 Supplier Abnova Corporation Supplier No. MAB7389
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant KITLG.

This gene encodes the ligand of the tyrosine-kinase receptor encoded by the KIT locus. This ligand is a pleiotropic factor that acts in utero in germ cell and neural cell development, and hematopoiesis, all believed to reflect a role in cell migration. In adults, it functions pleiotropically, while mostly noted for its continued requirement in hematopoiesis. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen KITLG
Applications ELISA
Classification Monoclonal
Clone four helix cytokine
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant KITLG.
Dilution ELISA (5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene KITLG
Gene Alias DKFZp686F2250/KL-1/Kitl/MGF/SCF/SF/SHEP7
Gene Symbols KITLG
Host Species Mouse
Immunogen Recombinant protein corresponding to human KITLG.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4254
Target Species Human, Rat
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.