Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF10 (M65J), Mouse anti-Human, Clone: 7C1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant KLF10.
Supplier: Abnova Corporation H00007071M65J
Description
Sequence: LSDTAKPHIAAPFKEEEKSPVSAPKLPKAQATSVIRHTADAQLCNHQTCPMKAASILNYQNNSFRRRTHLNVEAARKNIPCAAVSPNRSKCERNTVADVDSpecifications
KLF10 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC011538.1 | |
KLF10 | |
KLF10 (AAH11538.1,111 a.a. ∽ 210 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
7C1 | |
Mouse monoclonal antibody raised against a partial recombinant KLF10. | |
KLF10 | |
EGRA/TIEG/TIEG1 | |
Mouse | |
Affinity chromatography | |
RUO | |
7071 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction