Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-KLF15, Clone: 2G8,

Catalog No. MABS676MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS676MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS676MI Supplier MilliporeSigma™ Supplier No. MABS676
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects KLF15 clone: 2G8 in Human samples, and it is validated for

KLF15 (Kruppel-like factor 15), also known as KKLF, DKFZp779M1320. Entrez Protein NP_054798. Sequence analysis predicted that the 416 amino acid KLF15 protein, which is 84% identical to the rat Klf15 protein, contains 3 zinc finger motifs at its C terminus, N-terminal serine-rich stretches, and a central proline-rich segment.KLF15, a kidney-enriched Krueppel-like factor, is a transcriptonal activator that binds the CLCNKA promoter. KLF15, which is a nuclear protein, is expressed primarily in liver, heart, skeletal muscle and kidney tissues but is not detected in lymphoid tissues or bone marrow. KLF15 is an important regulator of GLUT4 in both adipose and muscle tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KLF15
Classification Monoclonal
Clone 2G8
Formulation Mouse monoclonal IgG1 ascitic fluid containing 0.03% sodium azide. 
Gene Accession No. Q9UIH9
Gene Symbols KLF15; KKLF
Host Species Mouse
Immunogen Purified recombinant fragment of human KLF15 expressed in E. coli.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.