Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF4, Mouse, Clone: 4E5C3, Abnova™
Mouse monoclonal antibody raised against KLF4.
Supplier: Abnova Corporation MAB11171
Specifications
KLF4 | |
Monoclonal | |
1 mg/mL | |
Mouse monoclonal antibody raised against KLF4. | |
In PBS (0.02% sodium azide) | |
EZF/GKLF | |
Mouse | |
Peptide affinity purification | |
RUO | |
9314 | |
Human, Mouse, Rat | |
Liquid |
ELISA, Western Blot | |
4E5C3 | |
Unconjugated | |
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
KLF4 | |
KLF4 | |
A synthetic peptide corresponding to 20 amino acids at C-terminus of human KLF4. | |
100 μg | |
Primary | |
At least three isoforms of KLF4 are known to exist; this antibody will detect all three. KLF4 antibody will not cross-react with other Kruppel-like family members. | |
Store at -20°C, stable for one year. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction