Learn More
KLF5, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant KLF5.
Supplier: Abnova Corporation H00000688A01
Description
This gene encodes a member of the Kruppel-like factor subfamily of zinc finger proteins. Since the protein localizes to the nucleus and binds the epidermal growth factor response element, it is thought to be a transcription factor. [provided by RefSeq
Sequence: RYNRRSNPDLEKRRIHYCDYPGCTKVYTKSSHLKAHLRTHTGEKPYKCTWEGCDWRFARSDELTRHYRKHTGAKPFQCGVCNRSFSRSDHLALHMKRHQNSpecifications
KLF5 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant KLF5. | |
KLF5 | |
BTEB2/CKLF/IKLF | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_001730 | |
KLF5 | |
KLF5 (NP_001721, 358 a.a. ∽ 457 a.a) partial recombinant protein with GST tag. | |
RUO | |
688 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only