Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLH, Mouse, Clone: 11E8F5C3, Abnova™

Catalog No. 89136594 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-594 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-594 Supplier Abnova Corporation Supplier No. MAB8544

Mouse monoclonal antibody raised against KLH .

Specifications

Antigen KLH
Applications ELISA, Western Blot
Classification Monoclonal
Clone 11E8F5C3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against KLH .
Dilution ELISA (1:2000-1:5000) Western Blot (2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS, pH 7.5, 0.1% BSA
Host Species Mouse
Immunogen Keyhole Limpet Hemocyanin, KLH.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody detected specifically individual KLH and conjugated protein, as well as KLH-conjugated peptide in both ELISA and Western Blot.
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.