Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLK3, Mouse, HRP, Clone: 5A6, Abnova™

Catalog No. 89112263 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-263 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-263 Supplier Abnova Corporation Supplier No. MAB5475
Add to Cart
Add to Cart
Request a Quote

Mouse monoclonal antibody raised against native KLK3.

Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. Its protein product is a protease present in seminal plasma. It is thought to function normally in the liquefaction of seminal coagulum, presumably by hydrolysis of the high molecular mass seminal vesicle protein. Serum level of this protein, called PSA in the clinical setting, is useful in the diagnosis and monitoring of prostatic carcinoma. Alternate splicing of this gene generates several transcript variants encoding different isoforms. [provided by RefSeq

Specifications

Antigen KLK3
Applications ELISA
Classification Monoclonal
Clone 5A6
Conjugate HRP
Description Mouse monoclonal antibody raised against native KLK3.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 5% BSA, 0.05% proclin 300)
Gene KLK3
Gene Alias APS/KLK2A1/PSA/hK3
Gene Symbols KLK3
Host Species Mouse
Immunogen Native purified KLK3 from human seminal plasma.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 354
Test Specificity Specific for total PSA in equimolar determination, epitope 6. No cross-reactivity with human kallikrein 2.
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.