Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLKB1, Mouse, Clone: 901, Abnova™

Catalog No. 89111991 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-991 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-991 Supplier Abnova Corporation Supplier No. MAB4873
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native KLKB1 (Heavy Chain).

Plasma prekallikrein is a glycoprotein that participates in the surface-dependent activation of blood coagulation, fibrinolysis, kinin generation and inflammation. It is synthesized in the liver and secreted into the blood as a single polypeptide chain. Plasma prekallikrein is converted to plasma kallikrein by factor XIIa by the cleavage of an internal Arg-Ile bond. Plasma kallikrein therefore is composed of a heavy chain and a light chain held together by a disulphide bond. The heavy chain originates from the amino-terminal end of the zymogen and contains 4 tandem repeats of 90 or 91 amino acids. Each repeat harbors a novel structure called the apple domain. The heavy chain is required for the surface-dependent pro-coagulant activity of plasma kallikrein. The light chain contains the active site or catalytic domain of the enzyme and is homologous to the trypsin family of serine proteases. Plasma prekallikrein deficiency causes a prolonged activated partial thromboplastin time in patients. [provided by RefSeq

Specifications

Antigen KLKB1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 901
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native KLKB1 (Heavy Chain).
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene KLKB1
Gene Alias KLK3/PPK
Gene Symbols KLKB1
Host Species Mouse
Immunogen Native purified human plasma KLKB1 (Heavy Chain).
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3818
Test Specificity Recognizes two variants (88 KDa and 85 KDa) of prekallikrein and its activation products kallikrein (88 KDa and 85 KDa), the complexes formed by kallikrein with its endogenous inhibitors C1 inhibitor, alpha-2-macroglobulin and Antithrombin III, and 45 KDa prekallikrein/kallikrein fragment(s). Recognizes prekallikrein and its activation products in human, chimpanzee, rhesus, and baboon plasmas. The epitope for this antibody, located on the prekallikrein/kallikrein heavy chain, is involved in the interaction between prekallikrein and factor XIIa. This antibody inhibits prekallikrein activation in human and rhesus plasmas by approximately 60-80% and 55%, respectively. This antibody does not cross-react with tissue kallikrein.
Target Species Baboon, Chimpanzee, Human, Monkey
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.