Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Klra6, Mouse, Clone: HBF-719, Abnova™

Catalog No. 89111290 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-290 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-290 Supplier Abnova Corporation Supplier No. MAB3242
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Klra6.

Specifications

Antigen Klra6
Applications Flow Cytometry
Classification Monoclonal
Clone HBF-719
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Klra6.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Klra6
Gene Alias Ly49f
Gene Symbols Klra6
Host Species Mouse
Immunogen Native mouse Klra6.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16637
Test Specificity B6 allele of the Ly49F receptor.
Target Species Mouse
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.