Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Klra6, Mouse, FITC, Clone: HBF-719, Abnova™

Catalog No. 89112440 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-440 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-440 Supplier Abnova Corporation Supplier No. MAB5769
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Klra6.

Specifications

Antigen Klra6
Applications Flow Cytometry
Classification Monoclonal
Clone HBF-719
Conjugate FITC
Description Mouse monoclonal antibody raised against Klra6.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Klra6
Gene Alias Ly49f
Gene Symbols Klra6
Host Species Mouse
Immunogen CHO cells transfected with the B6 allele of Ly49F.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16637
Test Specificity B6 allele of the Ly49F receptor.
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.