Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Klrb1c, Mouse, FITC, Clone: PK136, Abnova™

Catalog No. 89112446 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-446 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-446 Supplier Abnova Corporation Supplier No. MAB5775
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native Klrb1c.

Specifications

Antigen Klrb1c
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone PK136
Conjugate FITC
Description Mouse monoclonal antibody raised against native Klrb1c.
Dilution Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene Klrb1c
Gene Alias AI462337/CD161/Ly-59/Ly55c/Ly59/NK-RP1/NK1.1/NKRP1/NKRP140/Nk-1/Nk-1.2/Nk1/Nk1.2/Nkrp1c
Gene Symbols Klrb1c
Host Species Mouse
Immunogen Native purified Klrb1c from CE mouse spleen and bone marrow cells enriched for NK-1+ cells.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17059
Test Specificity Mouse NK1.1 alloantigen, Mr 80-85 KDa (39 KDa under reducing conditions).
Target Species Mouse
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.