Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KREMEN2, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107324 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-324 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-324 Supplier Abnova Corporation Supplier No. H00079412A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant KREMEN2.

This gene encodes a high-affinity dickkopf homolog 1 (DKK1) transmembrane receptor that functionally cooperates with DKK1 to block wingless (WNT)/beta-catenin signaling. The encoded protein forms a ternary membrane complex with DKK1 and the WNT receptor lipoprotein receptor-related protein 6 (LRP6), and induces rapid endocytosis and removal of LRP6 from the plasma membrane. It contains extracellular kringle, WSC, and CUB domains. Alternatively spliced transcript variants encoding distinct isoforms have been observed for this gene. [provided by RefSeq

Sequence: MPGYLGCFVDSGAPPALSGPSGTSTKLTVQVCLRFCRMKGYQLAGVEAGYACFCGSESDLARGRLAPATDCDQICFGHPGQLCGGDGRLGVYEVSVGSCQ

Specifications

Antigen KREMEN2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant KREMEN2.
Formulation 50% glycerol
Gene KREMEN2
Gene Accession No. NM_024507
Gene Alias KRM2/MGC10791/MGC16709
Gene Symbols KREMEN2
Host Species Mouse
Immunogen KREMEN2 (NP_078783, 121 a.a. ∽ 220 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79412
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.