Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT10, Mouse, Clone: LH2, Abnova™

Catalog No. 89109524 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-524 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-524 Supplier Abnova Corporation Supplier No. MAB11362
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against KRT10.

This gene encodes a member of the type I (acidic) cytokeratin family, which belongs to the superfamily of intermediate filament (IF) proteins. Keratins are heteropolymeric structural proteins which form the intermediate filament. These filaments, along with actin microfilaments and microtubules, compose the cytoskeleton of epithelial cells. Mutations in this gene are associated with epidermolytic hyperkeratosis. This gene is located within a cluster of keratin family members on chromosome 17q21. [provided by RefSeq

Specifications

Antigen KRT10
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LH2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against KRT10.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (0.5-1 ug/mL for 30 min at RT) Immunoprecipitation (0.5-1 ug/500 ug protein lysate) Western Blot (0.25-0.5 ug/mL) The optimal working dilution sho
Formulation In PBS, pH 7.4 (0.05% BSA, 0.05% sodium azide)
Gene KRT10
Gene Alias CK10/K10/KPP
Gene Symbols KRT10
Host Species Mouse
Immunogen Skin extract of a human Psoriasis patient.
Purification Method Protein A/G affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3858
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.