Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT10/KRT13, Mouse, Clone: CK 208, Abnova™

Catalog No. 89112729 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-729 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-729 Supplier Abnova Corporation Supplier No. MAB6192
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native KRT10/KRT13.

Specifications

Antigen KRT10/KRT13
Applications Immunocytochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone CK 208
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native KRT10/KRT13.
Dilution Immunohistochemistry (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (1% BSA, 0.05% sodium azide)
Gene Alias CK10/K10/KPP
Gene Symbols KRT10
Host Species Mouse
Immunogen Native purified human KRT10/KRT13.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3858/3860
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.