Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT14, Mouse, Clone: LL002, Abnova™

Catalog No. 89109500 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-500 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-500 Supplier Abnova Corporation Supplier No. MAB11337
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of KRT14.

This gene encodes a member of the keratin family, the most diverse group of intermediate filaments. This gene product, a type I keratin, is usually found as a heterotetramer with two keratin 5 molecules, a type II keratin. Together they form the cytoskeleton of epithelial cells. Mutations in the genes for these keratins are associated with epidermolysis bullosa simplex. At least one pseudogene has been identified at 17p12-p11. [provided by RefSeq

Specifications

Antigen KRT14
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LL002
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of KRT14.
Dilution Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (1-2 ug/mL for 30 min at RT) Immunoprecipitation (1-2 ug/500 ug protein) Western Blot (0.25-0.5 ug/mL) The optimal working dilution should be determ
Formulation In PBS, pH 7.4 (0.05% BSA, 0.05% sodium azide)
Gene KRT14
Gene Alias CK14/EBS3/EBS4/K14/NFJ
Gene Symbols KRT14
Host Species Mouse
Immunogen A synthetic peptide corresponding to 15 residues from the C-terminus of human KRT14.
Purification Method Protein A/G affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3861
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.