Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT18, Mouse, Clone: SPM265, Abnova™

Catalog No. 89111059 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-059 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-059 Supplier Abnova Corporation Supplier No. MAB2540
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native KRT18.

KRT18 encodes the type I intermediate filament chain keratin 18. Keratin 18, together with its filament partner keratin 8, are perhaps the most commonly found members of the intermediate filament gene family. They are expressed in single layer epithelial tissues of the body. Mutations in this gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Specifications

Antigen KRT18
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone SPM265
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native KRT18.
Dilution Western Blot (0.1-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50: 2-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In TBS, pH 7.2 (BSA, 10% Proclin300)
Gene KRT18
Gene Alias CYK18/K18
Gene Symbols KRT18
Host Species Mouse
Immunogen Native KRT18 from human breast cancer PMC 42 cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3875
Test Specificity This antibody recognizes human KRT18,
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.