Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRT19, Mouse, Biotin, Clone: A53-B/A2, Abnova™

Catalog No. 89112909 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-909 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-909 Supplier Abnova Corporation Supplier No. MAB6502
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native KRT19.

The protein encoded by this gene is a member of the keratin family. The keratins are intermediate filament proteins responsible for the structural integrity of epithelial cells and are subdivided into cytokeratins and hair keratins. The type I cytokeratins consist of acidic proteins which are arranged in pairs of heterotypic keratin chains. Unlike its related family members, this smallest known acidic cytokeratin is not paired with a basic cytokeratin in epithelial cells. It is specifically expressed in the periderm, the transiently superficial layer that envelopes the developing epidermis. The type I cytokeratins are clustered in a region of chromosome 17q12-q21. [provided by RefSeq

Specifications

Antigen KRT19
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A53-B/A2
Concentration 1 mg/mL
Conjugate Biotin
Description Mouse monoclonal antibody raised against native KRT19.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, 300mM NaCl, pH 7.4
Gene KRT19
Gene Alias CK19/K19/K1CS/MGC15366
Gene Symbols KRT19
Host Species Mouse
Immunogen Native purified KRT19 from human breast adenocarcinoma cell line (MCF-7).
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3880
Test Specificity This antibody reacts with Rod domain of cytokeratin 19 (40 KDa) in human tissue. Cytokeratin 19 is not expressed in hepatocytes; it is often co-expressed with cytokeratin 7.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.