Learn More
KRT5, Mouse, Clone: 7H8C4, Abnova™
Mouse monoclonal antibody raised against partial recombinant KRT5.
Supplier: Abnova Corporation MAB10727
Description
The protein encoded by this gene is a member of the keratin gene family. The type II cytokeratins consist of basic or neutral proteins which are arranged in pairs of heterotypic keratin chains coexpressed during differentiation of simple and stratified epithelial tissues. This type II cytokeratin is specifically expressed in the basal layer of the epidermis with family member KRT14. Mutations in these genes have been associated with a complex of diseases termed epidermolysis bullosa simplex. The type II cytokeratins are clustered in a region of chromosome 12q12-q13. [provided by RefSeq
Specifications
| KRT5 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| KRT5 | |
| KRT5 | |
| Recombinant protein corresponding to human KRT5. | |
| RUO | |
| 3852 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 7H8C4 | |
| Mouse monoclonal antibody raised against partial recombinant KRT5. | |
| In ascites (0.03% sodium azide) | |
| CK5/DDD/EBS2/K5/KRT5A | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.