Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KSR2, Mouse, Clone: 6F12, Abnova™

Catalog No. 89107746 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-746 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-746 Supplier Abnova Corporation Supplier No. H00283455M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant KSR2.

Sequence: PAPPLPPSATPPSPLHPSPQCTRQQKNFNLPASHYYKYKQQFIFPDVVPVPETPTRAPQVILHPVTSNPILEGNPLLQIEVEPTSENEEV

Specifications

Antigen KSR2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6F12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant KSR2.
Formulation ascites with no preservative
Gene KSR2
Gene Accession No. NM_173598
Gene Alias FLJ25965
Gene Symbols KSR2
Host Species Mouse
Immunogen KSR2 (NP_775869,411 a.a. ∽ 500 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283455
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.