Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse anti-Ku70/XRCC6, Clone: KU729, Azide Free, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
| Antigen | Ku70/XRCC6 |
|---|---|
| Clone | KU729 |
| Concentration | 1.0 mg/mL |
| Dilution | Immunocytochemistry/Immunofluorescence 0.5-1.0ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate |
| Applications | Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP23466302
![]() |
Novus Biologicals
NBP2346630.2 MG |
0.2 mg | N/A | N/A | N/A | ||||
Description
Ku70/XRCC6 Monoclonal antibody specifically detects Ku70/XRCC6 in Human, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative) samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Ku70/XRCC6 | |
| 1.0 mg/mL | |
| Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, DNA Double Strand Break Repair, DNA Repair, Non homologous end joining | |
| 10mM PBS - BSA free with No Preservative | |
| 5'-deoxyribose-5-phosphate lyase Ku70, 5'-dRP lyase Ku70, 70 kDa subunit of Ku antigen, ATP-dependent DNA helicase 2 subunit 1, ATP-dependent DNA helicase II 70 kDa subunit, CTC box binding factor 75 kDa subunit, CTC box-binding factor 75 kDa subunit, CTC75, CTCBF, D22S731, EC 3.6.4.-, EC 4.2.99.-, G22P1Ku70, Ku autoantigen p70 subunit, Ku autoantigen, 70kDa, KU70, ML8,70 kDa subunit, Thyroid-lupus autoantigen, thyroid-lupus autoantigen p70, TLAAD22S671, X-ray repair complementing defective repair in Chinese hamster cells 6DNA repair protein XRCC6, X-ray repair cross-complementing protein 6 | |
| XRCC6 | |
| IgG1 κ | |
| Protein A purified | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 75 kDa |
| KU729 | |
| Immunocytochemistry/Immunofluorescence 0.5-1.0ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Primate, Chicken (Negative), Mouse (Negative), Rat (Negative) | |
| P12956 | |
| 2547 | |
| Nuclear extract of human HL-60 cells | |
| Primary | |
| Protein A purified | |
| Recognizes a dimer of two proteins of 70kDa and ∼80kDa, identified as two subunits of Ku. This MAb recognizes a conformational epitope of p70/p80 dimer, which is destroyed during Western blotting. The p70/p80 dimer is important for function of a 460kDa DNA-dependent protein kinase. Ku protein plays a role in cell signaling, proliferation, DNA repair, replication, transcriptional activation, and apoptosis. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title