Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

La Crosse virus, Mouse, Clone: 9E7.2, Abnova™

Catalog No. 89136435 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-136-435 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-136-435 Supplier Abnova Corporation Supplier No. MAB8234
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against La Crosse virus.

Specifications

Antigen La Crosse virus
Applications ELISA, Immunofluorescence, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9E7.2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against La Crosse virus.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Mouse
Immunogen Native La Crosse Virus (LACV) from E6 Vero cells.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity G2 envelope glycoprotein (35kD); virus-neutralizing
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.