Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-LAG3, Clone: 8C7.1,

Catalog No. MABF294MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF294MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF294MI Supplier MilliporeSigma™ Supplier No. MABF294
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects LAG3 clone: 8C7.1 in Human samples, and it is validated for Flow Cytometry, Western Blotting

Lymphocyte activation gene 3 protein (UniProt P18627; also known as CD223, LAG-3, Protein FDC) is encoded by the LAG3 (also known as FDC) gene (Gene ID 3902) in human. LAG3 is a single-pass type I membrane protein that binds MHC class II molecules on antigen presenting cells with high affinity. LAG3 expression is induced after T cell activation and functions as a negative regulator of T cell function. LAG3 blockage is shown to augment T cell proliferation and cytokine production, and LAG3-deficient mice have a defect in the downregulation of T cell responses induced by staphylococcal enterotoxin B or by virus infection. In addition, LAG3 is expressed on activated regulatory T (Treg) cells at higher levels than on activated effector T cells. LAG3 blockage inhibits the suppressive activity of Treg cells. LAG3 is also upregulated on tumour-infiltrating lymphocytes (TILs), and LAG3 blockage can enhance anti-tumour T cell responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAG3
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone 8C7.1
Formulation Mouse monoclonal IgG2aκ antibody tissue culture supernatant with 0.05% sodium azide.
Gene Accession No. P18627
Gene Symbols LAG3; FDC
Host Species Mouse
Immunogen Mouse Fc-tagged recombinant human LAG3 extracellular fragment.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Inflammation and Immunology
Primary or Secondary Primary
Gene ID (Entrez) NP_002277
Target Species Human
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Supernatant
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.