Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LALBA, Mouse, Clone: F20.16, Abnova™

Catalog No. 89113148 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-148 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-148 Supplier Abnova Corporation Supplier No. MAB7211
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against LALBA.

This gene encodes alpha-lactalbumin, a principal protein of milk. Alpha-lactalbumin forms the regulatory subunit of the lactose synthase (LS) heterodimer and beta 1,4-galactosyltransferase (beta4Gal-T1) forms the catalytic component. Together, these proteins enable LS to produce lactose by transferring galactose moieties to glucose. As a monomer, alpha-lactalbumin strongly binds calcium and zinc ions and may possess bactericidal or antitumor activity. A folding variant of alpha-lactalbumin, called HAMLET, likely induces apoptosis in tumor and immature cells. [provided by RefSeq

Specifications

Antigen LALBA
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone F20.16
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against LALBA.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene LALBA
Gene Alias MGC138521/MGC138523
Gene Symbols LALBA
Host Species Mouse
Immunogen Human LALBA.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3906
Test Specificity alpha Lactalbumin (F20.16). Detects a band of approximately 14 KDa.
Target Species Human
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.