Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMA3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104823 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-823 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-823 Supplier Abnova Corporation Supplier No. H00003909A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant LAMA3.

Laminins are basement membrane components thought to mediate the attachment, migration and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components. The protein encoded by this gene is the alpha-3 subunit of laminin 5, which is a complex glycoprotein composed of three subunits (alpha, beta, and gamma). Laminin 5 is thought to be involved in cell adhesion, signal transduction and differentiation of keratinocytes. Mutations in this gene have been identified as the cause of Herlitz type junctional epidermolysis bullosa. Alternatively spliced transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq

Sequence: SSQQQRVPFLQPPGQSQLQASYVEFRPSQGCSPGYYRDHKGLYTGRCVPCNCNGHSNQCQDGSGICVNCQHNTAGEHCERCQEGYYGNAVHGSCRACPCPHTNSFATGCV

Specifications

Antigen LAMA3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant LAMA3.
Formulation 50% glycerol
Gene LAMA3
Gene Accession No. NM_000227
Gene Alias BM600/E170/LAMNA/LOCS/lama3a
Gene Symbols LAMA3
Host Species Mouse
Immunogen LAMA3 (NP_000218, 29 a.a. ∽ 138 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3909
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.