Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-LAMA4, Clone: 9B2, Abnova™

Catalog No. 89113198 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-198 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-198 Supplier Abnova Corporation Supplier No. MAB7871

Specifications

Antigen LAMA4
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 9B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native LAMA4.
Dilution ELISA (0.125 ug/mL) Western Blot (0.5-1 ug/mL) Immunoprecipitation (0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene LAMA4
Gene Alias DKFZp686D23145/LAMA3/LAMA4*-1
Gene Symbols LAMA4
Host Species Mouse
Immunogen Immunoaffinity-purified Laminin 411 (laminin-8 (alpha4beta1gamma1) from platelets.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3910
Target Species Human
Content And Storage Store at -20°C. For long term storage store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.