Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-LAMB1, Clone: 2D9G5, Abnova™

Catalog No. 89108752 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-752 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-752 Supplier Abnova Corporation Supplier No. MAB10561
Add to Cart
Add to Cart

Specifications

Antigen LAMB1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2D9G5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant LAMB1.
Dilution ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene LAMB1
Gene Alias CLM/MGC142015
Gene Symbols LAMB1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 31-270 of human LAMB1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3912
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.