Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-LAMC1, Clone: 39, Abnova™

Catalog No. 89113861 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-861 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-861 Supplier Abnova Corporation Supplier No. MAB9903
Add to Cart
Add to Cart

Specifications

Antigen LAMC1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 39
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant LAMC1.
Dilution Western blot (1:1000) The optimal working dilution should be determined by the end user.
Formulation In Citrate-Tris-HCl buffer, pH 7.0 (0.02% Proclin 300)
Gene LAMC1
Gene Accession No. NM_002293
Gene Alias LAMB2/MGC87297
Gene Symbols LAMC1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 526-1106 of human LAMC1.
Purification Method Affinity purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3915
Test Specificity It can expression in SW480 whole cell lysate.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.