Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-LAMC1, Clone: BC17, Abnova™

Catalog No. 89111052 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-052 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-052 Supplier Abnova Corporation Supplier No. MAB2519
Add to Cart
Add to Cart

Specifications

Antigen LAMC1
Applications Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone BC17
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial native LAMC1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (1% BSA, 0.09% sodium azide)
Gene LAMC1
Gene Alias LAMB2/MGC87297
Gene Symbols LAMC1
Host Species Mouse
Immunogen Native purified human LAMC1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3915
Test Specificity This antibody recognizes the Mr 200,000 gamma1-chain of human reduced and alkylated laminin produced by various cultured cells, which produce laminins complexed with it. The specificity of Mab BC17 has been confirmed by immunoprecipitation and Elisa by using recombinant gamma1- chain. This antibody only reacts with human cells and tissues. The monoclonal antibody can be widely applied in immunohistochemical studies as all basement membranes contain laminin trimers containing these chains.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.