Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP1, Mouse, PE, Clone: H4A3, Abnova™

Catalog No. 89108442 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-442 100 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-442 Supplier Abnova Corporation Supplier No. MAB10237
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against of LAMP1.

The protein encoded by this gene is a member of a family of membrane glycoproteins. This glycoprotein provides selectins with carbohydrate ligands. It may also play a role in tumor cell metastasis. [provided by RefSeq

Specifications

Antigen LAMP1
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone H4A3
Conjugate PE
Description Mouse monoclonal antibody raised against of LAMP1.
Dilution Flow Cytometry (4 uL reagent/100 uL of whole blood or 106 cells) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.02% sodium azide)
Gene LAMP1
Gene Alias CD107a/LAMPA/LGP120
Gene Symbols LAMP1
Host Species Mouse
Immunogen Human PBMCs.
Purification Method Size-exclusion chromatography purification
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3916
Test Specificity LAMP1, a glycoprotein expressed mainly on lysosomal, but also on the plasma membrane.
Target Species Human, Mouse, Non-human Primate
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.