Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAMP2, Mouse, Clone: H4B4, Abnova™

Catalog No. 89108443 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-443 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-443 Supplier Abnova Corporation Supplier No. MAB10238
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against of LAMP2.

The protein encoded by this gene is a member of a family of membrane glycoproteins. This glycoprotein provides selectins with carbohydrate ligands. It may play a role in tumor cell metastasis. It may also function in the protection, maintenance, and adhesion of the lysosome. Alternative splicing of this gene results in multiple transcript variants encoding distinct proteins. [provided by RefSeq

Specifications

Antigen LAMP2
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone H4B4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against of LAMP2.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene LAMP2
Gene Alias CD107b/LAMPB/LGP110
Gene Symbols LAMP2
Host Species Mouse
Immunogen Human PBMCs.
Purification Method Protein A purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3920
Test Specificity LAMP2, an extensively glycosylated 100-120 kDa widely expressed lysosome-associated protein.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.