Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lat, Mouse, Clone: LAT-1D1, Abnova™

Catalog No. 89111503 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-503 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-503 Supplier Abnova Corporation Supplier No. MAB3623

Mouse monoclonal antibody raised against full-length recombinant Lat.

Specifications

Antigen Lat
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LAT-1D1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant Lat.
Dilution Western Blot (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene Lat
Gene Alias MGC108666
Gene Symbols Lat
Host Species Mouse
Immunogen Recombinant protein corresponding to full length rat Lat.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81511
Test Specificity This antibody reacts with mouse and rat Linker for activation of T cells (Lat), a transmembrane adaptor protein expressed by T cells, pre-B cells, NK cells, mast cells andplatelets.
Target Species Mouse, Rat
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.