Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LATS2, Mouse, Clone: ST-3B11, Abnova™

Catalog No. 89107820 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-820 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-820 Supplier Abnova Corporation Supplier No. MAB0009

Mouse monoclonal antibody raised against synthetic phosphopeptide of LATS2.

This gene encodes a serine/threonine protein kinase belonging to the LATS tumor suppressor family. The protein localizes to centrosomes during interphase, and early and late metaphase. It interacts with the centrosomal proteins aurora-A and ajuba and is required for accumulation of gamma-tubulin and spindle formation at the onset of mitosis. It also interacts with a negative regulator of p53 and may function in a positive feedback loop with p53 that responds to cytoskeleton damage. Additionally, it can function as a co-repressor of androgen-responsive gene expression. [provided by RefSeq

Specifications

Antigen LATS2
Applications ELISA, Western Blot
Classification Monoclonal
Clone ST-3B11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic phosphopeptide of LATS2.
Dilution Western Blot (1-2 ug/mL) ELISA (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM HEPES/KOH, 150mM NaCl, pH 7.5 (50% glycerol)
Gene LATS2
Gene Accession No. NM_014572
Gene Alias FLJ13161/KPM
Gene Symbols LATS2
Host Species Mouse
Immunogen Synthetic phosphopeptide (conjugated with KLH) corresponding to residues surrounding S83 of human LATS2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26524
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.