Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LBP, Mouse, Clone: biG 42, Abnova™

Catalog No. 89112352 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-352 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-352 Supplier Abnova Corporation Supplier No. MAB5661
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant LBP.

The protein encoded by this gene is involved in the acute-phase immunologic response to gram-negative bacterial infections. Gram-negative bacteria contain a glycolipid, lipopolysaccharide (LPS), on their outer cell wall. Together with bactericidal permeability-increasing protein (BPI), the encoded protein binds LPS and interacts with the CD14 receptor, probably playing a role in regulating LPS-dependent monocyte responses. Studies in mice suggest that the encoded protein is necessary for the rapid acute-phase response to LPS but not for the clearance of LPS from circulation. This protein is part of a family of structurally and functionally related proteins, including BPI, plasma cholesteryl ester transfer protein (CETP), and phospholipid transfer protein (PLTP). Finally, this gene is found on chromosome 20, immediately downstream of the BPI gene. [provided by RefSeq

Specifications

Antigen LBP
Applications ELISA, Western Blot
Classification Monoclonal
Clone biG 42
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant LBP.
Dilution ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS
Gene LBP
Gene Alias MGC22233
Gene Symbols LBP
Host Species Mouse
Immunogen Recombinant protein corresponding to human LBP.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3929
Test Specificity LPS binding protein (LBP) is an approximately 60 KDa acute phase protein that is produced by hepatocytes. This protein strongly binds to LPS and has been shown to play an important role in the handling of LPS by the host. A number of functions of LBP have been reported. First, LBP transfers LPS to the LPS receptor CD14 on mononuclear phagocytes, leading to a 100-1,000-fold increased sensitivity of the cells to LPS. Furthermore, LBP can enhance the response of CD14 negative cells by acceleration of LPS binding to soluble CD14, a complex that stimulates these cells. LBP was demonstrated to protect mice from septic shock caused by LPS or gram negative bacteria.
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.