Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCK, Mouse, Clone: 8E5F9, 4B2C7, Abnova™

Catalog No. 89112279 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-279 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-279 Supplier Abnova Corporation Supplier No. MAB5528
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant LCK.

This gene is a member of the Src family of protein tyrosine kinases (PTKs). The encoded protein is a key signaling molecule in the selection and maturation of developing T-cells. It contains N-terminal sites for myristylation and palmitylation, a PTK domain, and SH2 and SH3 domains which are involved in mediating protein-protein interactions with phosphotyrosine-containing and proline-rich motifs, respectively. The protein localizes to the plasma membrane and pericentrosomal vesicles, and binds to cell surface receptors, including CD4 and CD8, and other signaling molecules. Multiple alternatively spliced variants, encoding the same protein, have been described. [provided by RefSeq

Specifications

Antigen LCK
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4B2C7, 8E5F9
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant LCK.
Dilution Western Blot (1:500-1:2000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation In ascites
Gene LCK
Gene Alias YT16/p56lck/pp58lck
Gene Symbols LCK
Host Species Mouse
Immunogen Recombinant protein corresponding to human LCK.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3932
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.