Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDHA, Mouse, Clone: AT1A4, Abnova™

Catalog No. 89113521 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-521 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-521 Supplier Abnova Corporation Supplier No. MAB8700
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant LDHA.

The protein encoded by this gene catalyzes the conversion of L-lactate and NAD to pyruvate and NADH in the final step of anaerobic glycolysis. The protein is found predominantly in muscle tissue and belongs to the lactate dehydrogenase family. Mutations in this gene have been linked to exertional myoglobinuria. Multiple transcript variants encoding different isoforms have been found for this gene. The human genome contains several nontranscribed pseudogenes of this gene. [provided by RefSeq

Specifications

Antigen LDHA
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone AT1A4
Concentration 1 mg/mL
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant LDHA.
Dilution Western Blot (1:500-1:8000) Immunofluorescence (1:500-1:8000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene LDHA
Gene Alias LDH-M/LDH1/PIG19
Gene Symbols LDHA
Host Species Mouse
Immunogen Recombinant protein corresponding to full length of human LDHA.
Purification Method Protein G purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3939
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.