Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LDOC1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106633 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-633 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-633 Supplier Abnova Corporation Supplier No. H00023641A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant LDOC1.

The protein encoded by this gene contains a leucine zipper-like motif and a proline-rich region that shares marked similarity with an SH3-binding domain. The protein localizes to the nucleus and is down-regulated in some cancer cell lines. It is thought to regulate the transcriptional response mediated by the nuclear factor kappa B (NF-kappaB). The gene has been proposed as a tumor suppressor gene whose protein product may have an important role in the development and/or progression of some cancers. [provided by RefSeq

Sequence: LSIENSQLMEQLRLLVCERASLLRQVRPPSCPVPFPETFNGESSRLPEFIVQTASYMLVNENRFCNDAMKVAFLISLLTGEAEEWVVPYIEMDSPILGDYRA

Specifications

Antigen LDOC1
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant LDOC1.
Formulation 50% glycerol
Gene LDOC1
Gene Accession No. NM_012317
Gene Alias BCUR1/Mar7/Mart7
Gene Symbols LDOC1
Host Species Mouse
Immunogen LDOC1 (NP_036449, 19 a.a. ∽ 120 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23641
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.