Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LEP, Mouse, Clone: 1E9 1H6, Abnova™

Catalog No. 89111535 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-535 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-535 Supplier Abnova Corporation Supplier No. MAB3685
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant LEP.

This gene encodes a protein that is secreted by white adipocytes, and which plays a major role in the regulation of body weight. This protein, which acts through the leptin receptor, functions as part of a signaling pathway that can inhibit food intake and/or regulate energy expenditure to maintain constancy of the adipose mass. This protein also has several endocrine functions, and is involved in the regulation of immune and inflammatory responses, hematopoiesis, angiogenesis and wound healing. Mutations in this gene and/or its regulatory regions cause severe obesity, and morbid obesity with hypogonadism. This gene has also been linked to type 2 diabetes mellitus development. [provided by RefSeq

Specifications

Antigen LEP
Applications ELISA
Classification Monoclonal
Clone 1E9 1H6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant LEP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS
Gene LEP
Gene Alias FLJ94114/OB/OBS
Gene Symbols LEP
Host Species Mouse
Immunogen Recombinant protein corresponding to human LEP.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3952
Test Specificity This antibody recognizes natural and recombinant human LEP.
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.