Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LGALS1, Mouse, Clone: 1E8-1B2, Abnova™

Catalog No. 89104834 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-834 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-834 Supplier Abnova Corporation Supplier No. H00003956M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant LGALS1.

The galectins are a family of beta-galactoside-binding proteins implicated in modulating cell-cell and cell-matrix interactions. This gene product may act as an autocrine negative growth factor that regulates cell proliferation. [provided by RefSeq

Sequence: MACGLVASNLNLKPGECLRVRGEVAPDAKSFVLNLGKDSNNLCLHFNPRFNAHGDANTIVCNSKDGGAWGTEQREAVFPFQPGSVAEVCITFDQANLTVKLPDGYEFKFPNRLNLEAINYMAADGDFKIKCVAFD

Specifications

Antigen LGALS1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E8-1B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant LGALS1.
Formulation ascites with no preservative
Gene LGALS1
Gene Accession No. BC001693
Gene Alias DKFZp686E23103/GAL1/GBP
Gene Symbols LGALS1
Host Species Mouse
Immunogen LGALS1 (AAH01693,1 a.a. ∽ 135 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3956
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2b λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.