Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-LILRA5/CD85f, Alexa Fluor 594, Clone: 711828, R&D Systems™

Catalog No. FAB6754T025
Change view
Click to view available options
Quantity:
25 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6754T025 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6754T025 Supplier R&D Systems Supplier No. FAB6754T025
This item is not returnable. View return policy

Specifications

Antigen ILT11/LILRA5
Applications Flow Cytometry
Classification Monoclonal
Clone 711828
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. A6NI73
Gene Alias CD85, CD85 antigen-like family member F, CD85F, CD85f antigen, ILT11, ILT-11, ILT11CD85f, Immunoglobulin-like transcript 11, leukocyte Ig-like receptor 9, Leukocyte immunoglobulin-like receptor 9, leukocyte immunoglobulin-like receptor subfamily A member 5, leukocyte immunoglobulin-like receptor subfamily A member 5 soluble, leukocyte immunoglobulin-like receptor, subfamily A (with TM domain), member 5, leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), LILRB7, LIR9, LIR-9, LIR9immunoglobulin-like transcript 11 protein, member 7
Gene Symbols LILRA5
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human ILT11/LILRA5, Gly42-Arg268, Accession # A6NI73
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 353514
Test Specificity Detects human ILT11/LILRA5 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse ILT11/LILRA5, recombinant rat (rr) ILT11/LILRA5, rrLILRC2, recombinant human (rh) ILT4/CD85d, rhILT5/CD85a, or rhILT3/CD85k is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.