Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LILRB4, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89106439 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-439 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-439 Supplier Abnova Corporation Supplier No. H00011006A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant LILRB4.

This gene is a member of the leukocyte immunoglobulin-like receptor (LIR) family, which is found in a gene cluster at chromosomal region 19q13.4. The encoded protein belongs to the subfamily B class of LIR receptors which contain two or four extracellular immunoglobulin domains, a transmembrane domain, and two to four cytoplasmic immunoreceptor tyrosine-based inhibitory motifs (ITIMs). The receptor is expressed on immune cells where it binds to MHC class I molecules on antigen-presenting cells and transduces a negative signal that inhibits stimulation of an immune response. The receptor can also function in antigen capture and presentation. It is thought to control inflammatory responses and cytotoxicity to help focus the immune response and limit autoreactivity. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MIPTFTALLCLGLSLGPRTDMQAGPLPKPTLWAEPGSVISWGNSVTIWCQGTLEAREYRLDKEESPAPWDRQNPLEPKNKARFSIPSMTEDYAGRYRCYYRSPVGWSQPSDPLELVMTGAYSKPTLSALPSPLVTSGKSVTLLCQSRSPMDTFLLIKERAAHPLLHLRSEHGAQQHQAEFPMSPVTSVHGGTYRCFSSHGFSHYLLSHPSDPLELIVSGSLEGPRPSPTRSVSTAAGPEDQPLMPTGSVPHSGLRRHWEVLIGVLVVSILLLSLLLFLLLQHWRQGKHRTLAQRQADFQRPPGAAEPEPKDGGLQRRSSPAADVQGENFCAAVKNTQPEDGVEMDTRQSPHDEDPQAVTYAKVKHSRPRREMASPPSPLSGEFLDTKDRQAEEDRQMDTEAAASEAPQDVTYARLHSFTLRQKATEPPPSQEGASPAEPSVYATLAIH

Specifications

Antigen LILRB4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant LILRB4.
Formulation 50% glycerol
Gene LILRB4
Gene Accession No. BC026309
Gene Alias CD85K/HM18/ILT3/LILRB5/LIR-5/LIR5
Gene Symbols LILRB4
Host Species Mouse
Immunogen LILRB4 (AAH26309, 1 a.a. ∽ 448 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11006
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.