Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LIME1, Mouse, Clone: LIME-06, Abnova™

Catalog No. 89108288 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-288 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-288 Supplier Abnova Corporation Supplier No. MAB0960
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant LIME1.

LIME1 is a raft-associated transmembrane adaptor phosphoprotein that is preferentially expressed in hemopoietic cells, particularly T cells (Brdickova et al., 2003 [PubMed 14610046]; Hur et al., 2003 [PubMed 14610044]).[supplied by OMIM

Specifications

Antigen LIME1
Applications Immunoprecipitation
Classification Monoclonal
Clone LIME-06
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant LIME1.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene LIME1
Gene Alias FLJ20406/LIME/LP8067/dJ583P15.4
Gene Symbols LIME1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 141-295 of human LIME1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54923
Test Specificity This antibody was raised against intracellular fragment corresponding to aa 141-295 of human LIME, a 31 KDa Lck-interacting transmembrane adaptor expressed by T cells.
Target Species Human
Content And Storage Store at 4°C. Do not freeze.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.