Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMNA, Mouse, Clone: 133A2, Abnova™

Catalog No. 89111018 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-018 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-018 Supplier Abnova Corporation Supplier No. MAB2470

Mouse monoclonal antibody raised against recombinant LMNA.

The nuclear lamina consists of a two-dimensional matrix of proteins located next to the inner nuclear membrane. The lamin family of proteins make up the matrix and are highly conserved in evolution. During mitosis, the lamina matrix is reversibly disassembled as the lamin proteins are phosphorylated. Lamin proteins are thought to be involved in nuclear stability, chromatin structure and gene expression. Vertebrate lamins consist of two types, A and B. Through alternate splicing, this gene encodes three type A lamin isoforms. Mutations in this gene lead to several diseases: Emery-Dreifuss muscular dystrophy, familial partial lipodystrophy, limb girdle muscular dystrophy, dilated cardiomyopathy, Charcot-Marie-Tooth disease, and Hutchinson-Gilford progeria syndrome. [provided by RefSeq

Specifications

Antigen LMNA
Applications Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 133A2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant LMNA.
Dilution Flow Cytometry (1:100-1:200) Western Blot (1:100-1:1000) Immunohistochemistry (1:100-1:200) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene LMNA
Gene Alias CDCD1/CDDC/CMD1A/CMT2B1/EMD2/FPL/FPLD/HGPS/IDC/LDP1/LFP/LGMD1B/LMN1/LMNC/PRO1
Gene Symbols LMNA
Host Species Mouse
Immunogen Recombinant protein corresponding to human LMNA.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4000
Test Specificity Recognizes an epitope located between residues 598-611 of LMNA, therefore it reacts exclusively with Lamin A.
Target Species Bovine, Canine, Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.