Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LMNB1, Mouse, Clone: 4B10, Abnova™

Catalog No. 89104846 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-846 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-846 Supplier Abnova Corporation Supplier No. H00004001M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant LMNB1.

The nuclear lamina consists of a two-dimensional matrix of proteins located next to the inner nuclear membrane. The lamin family of proteins make up the matrix and are highly conserved in evolution. During mitosis, the lamina matrix is reversibly disassembled as the lamin proteins are phosphorylated. Lamin proteins are thought to be involved in nuclear stability, chromatin structure and gene expression. Vertebrate lamins consist of two types, A and B. This gene encodes one of the two B type proteins, B1. [provided by RefSeq

Sequence: LGKCKAEHDQLLLNYAKKESDLNGAQIKLREYEAALNSKDAALATALGDKKSLEGDLEDLKDQIAQLEASLAAAKKQLAD

Specifications

Antigen LMNB1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant LMNB1.
Formulation ascites with no preservative
Gene LMNB1
Gene Accession No. NM_005573
Gene Alias ADLD/LMN/LMN2/LMNB/MGC111419
Gene Symbols LMNB1
Host Species Mouse
Immunogen LMNB1 (NP_005564.1,107 a.a. ∽ 186 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4001
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.